Adapter kit, Neuros™
供应商: HAMILTON BONADUZ
65460-01
HAMI65460-01EA 0 CNY
HAMI65460-01
Adapter kit, Neuros™
色谱用品 Chromatography Spare Parts
The Neuros™ adapter kit attaches to any 5,0 to 100 µl removable needle (RN) syringe.
  • Gas sterilisable, not autoclavable
  • Stainless steel, borosilicate glass fluid path
The needle sleeves come in two versions, one with a blind stop and one without. The sleeve with a stop is designed for applications where it is desirable to limit the penetration depth of the needle, as is the case with a cannulated animal. The sleeve without a stop supports the needle but does not obstruct the view of the injection target which is ideal for microsurgeries using aMore Product Information