Needle cleaning concentrate and wires
供应商: HAMILTON BONADUZ
18311 76620 18310 18300 18301 18302 18303 18304 18306
HAMI18311EA 0 CNY
HAMI18311 HAMI76620 HAMI18310 HAMI18300 HAMI18301 HAMI18302 HAMI18303 HAMI18304 HAMI18306
Needle cleaning concentrate and wires
清洗剂 洗涤剂和表面清洁剂
To clean syringes it is best to use a solvent known to be effective in solvating. Ideally the solvent would be non-alkaline, non-phosphate, and non-detergent based.
  • Biodegradable, non-phosphate, cleaning concentrate that is suitable for many common sample types
  • For some needle obstructions, mechanical cleaning may be necessary
For this purpose various sized tungsten cleaning wires can be used. They can be threaded through the needle to clear a blockage.
To clean syringes it is best to use a solvent known to be effective in solvating. Ideally theMore Product Information