Presence-Absence media in bottles
供应商: Hach
2401612
HACH2401612EA 0 CNY
HACH2401612
Presence-Absence media in bottles
微生物学媒介 准备微生物学媒介
Selective for Total Coliform Bacteria and Escherichia coli. Broth in pre-filled bottles.
  • Sterile
  • Disposable