Half area microplates, 96-well
供应商: GREINER BIO ONE
675076_40
GREI675076_40EA 0 CNY
GREI675076_40
Half area microplates, 96-well
微孔板 高温超导微孔板
PS, without lid. 96-well half area microplates offer an alternative to 384-well or 1536-well microplates when low sample volume is an important criterion but automation is not necessarily appropriate. They can be pipetted manually while at the same time allow a reduction in sample volume of up to 50%.
Microplates with clear bottoms feature micro-thin µClear® bottoms for exceptional optical properties and standardised pathlength, eliminating the need for pathlength determinations. White plates are ideal for luminescence assays, while black plates are suitable for fluorescence assays. BothMore Product Information