Microplates, 96-well
供应商: GREINER BIO ONE
650101_100 655101_100 655077_40 655075_40 655076_40 655074_40 655096_40
GREI650101_100EA 0 CNY
GREI650101_100 GREI655101_100 GREI655077_40 GREI655075_40 GREI655076_40 GREI655074_40 GREI655096_40
Microplates, 96-well
微孔板 高温超导微孔板
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
现在订购
规格
Environmentally Preferable Low Manufacturing Impact
环境认证 ISO 14001 (241002); ISO 50001 (241002)