Accessories for JB Academy, JB Nova, SUB Aqua Pro, SUB Aqua Plus, JB Aqua Plus and SBB Aqua Plus baths
供应商: GRANT INSTRUMENTS
LF14 LF28 LF36 LF6 LU14 LU28 LU36 LU6 RS14H RS28H RS36H SY1 J2-10 J2-13 J2-16 J2-19 J2-25 J2-30 J2-SE J2-LE AQBT2 AQBT26 AQBT5 AQL2 AQL5 AQBT12 RS18H
GRANLF14EA 0 CNY
GRANLF14 GRANLF28 GRANLF36 GRANLF6 GRANLU14 GRANLU28 GRANLU36 GRANLU6 GRANRS14H GRANRS28H GRANRS36H GRANSY1 GRANJ2-10 GRANJ2-13 GRANJ2-16 GRANJ2-19 GRANJ2-25 GRANJ2-30 GRANJ2-SE GRANJ2-LE GRANAQBT2 GRANAQBT26 GRANAQBT5 GRANAQL2 GRANAQL5 GRANAQBT12 GRANRS18H
Accessories for JB Academy, JB Nova, SUB Aqua Pro, SUB Aqua Plus, JB Aqua Plus and SBB Aqua Plus baths
浴锅 非搅拌水浴锅
A range of different stainless steel accessories for use with JB Academy, JB Nova, SUB Aqua Pro, (also previous models: SUB Aqua Plus, JB Aqua Plus and SBB Aqua Plus baths).
  • Flat lids with ring sets allow vessels of various sizes and heights to be used in the bath
  • Sloping lids, with insulated handle, direct condensation down the side of the bath, protecting vessels in the bath
  • Clear PC lids are supplied as standard with JB Nova and SUB Aqua Pro baths
  • Raised half shelves help accommodate different sizes of vessel simultaneously by altering the effective
More Product Information