Ultra-low temperature upright freezers, model SU780XLE
供应商: GLOBAL COOLING
SU780XLE
GLCLSU780XLEEA 0 CNY
GLCLSU780XLE
Ultra-low temperature upright freezers, model SU780XLE
冷柜 超低温冰箱
Ultra-low temperature freezer featuring the free-piston Stirling engine. This next generation of environmentally sustainable ULT freezers achieve stable storage conditions over a wide temperature range. The freezer uses 70 to 75% less energy than standard compressor-based ULT freezers, with the industry's best ultra-low cooling performance.
  • ±1 °C 'steady state' temperature variation over time
  • Faster initial pull down, ambient to –80 °C, <6,5 hours
  • Faster door opening temperature recovery, 35 minutes to –80 °C (ENERGY STAR® final test method)
  • Slowest
More Product Information