illustra™ ExoProStar™ kits

供应商: Cytiva

US78210 US78211 US78212 US78225 US78220
GEHEUS78210EA 0 CNY
GEHEUS78210 GEHEUS78211 GEHEUS78212 GEHEUS78225 GEHEUS78220
illustra™ ExoProStar™ kits
核酸试剂 核酸纯化试剂盒和试剂

illustra™ ExoProStar™ contains illustra™ Alkaline Phosphatase and Exonuclease 1, formulated to work together to remove unincorporated primers and nucleotides from amplification reactions in preparation for sequencing, cloning, genotyping or further DNA modification reactions.


  • Enzymes optimised to work together for high efficiency removal of unincorporated primers and nucleotides
  • Enzymes provided in two separate tubes, just two simple pipetting steps are needed to prepare the reaction
  • Fast 30 minute protocol; complete heat inactivation of the enzymes within 15 minutes
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Easy to automate

现在订购

规格

说明 illustra™ ExoProStar™ Enzymatic PCR and Sequence Reaction Clean Up Kit
Environmentally Preferable
Application
Target molecule DNA
Refine target molecule Amplified/modified DNA
Sample type PCR and other enzymatic reactions
Method / Format Enzymatic clean-up
Prep size Scalable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR