Anti-BrdU Mouse Monoclonal Antibody
供应商: GE HEALTHCARE
RPN202
GEHERPN202EA 0 CNY
GEHERPN202
Anti-BrdU Mouse Monoclonal Antibody
抗体
Anti-Bromodeoxyuridine (Anti-BRdU) antibody is supplied in the same formulation as the monoclonal antibody to BrdU in the Cell Proliferation Kit. The antibody/nuclease mixture is compatible with detection systems that incorporate fluorescently labelled antibodies and biotin-streptavidin detection reagents.
  • No need for chemical denaturation
  • Detection while maintaining cell morphology
  • Double immunocytochemical staining studies can be perfromed
Used in conjunction with RPN20, the reagent allows extended use of the kit where additional amounts of labelingMore Product Information
现在订购
规格
抗原 Bromodeoxyuridine,BrdU
抗原 Bromodeoxyuridine,BrdU
Environmentally Preferable
抗体类型 Primary
克隆形成能力 Monoclonal
结合 Unconjugated
反应性
主机 Mouse
同位型
免疫组织化学 Yes
荧光免疫检验法 Yes
存储温度 Cool
装运温度 Ice