Reactive dye 5-packs, Amersham CyDye™
供应商: Cytiva
PA23001
PA23500
PA25001
PA25500
GEHEPA23001EA
0
CNY
GEHEPA23001
GEHEPA23500
GEHEPA25001
GEHEPA25500
Reactive dye 5-packs, Amersham CyDye™
蛋白连接和标记系统
Mono-reactive dye pack contains derivatised CyDye™ that carries only one reactive group on each dye molecule for accurate labelling of amine groups, for example on antibodies or oligonucleotides.
- Convenient way to label antibodies, proteins, drugs, and oligonucleotides with CyDye™ fluorescent dyes
- For use in ELISA, immunoprecipitation, and blotting applications
- Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labellings
Bis-reactive dye pack contains fluors carrying two reactive groups on each dye molecule for general labelling of protein amine groups.
Delivery information: Each kit contains five vials of dye. Each vial is sufficient for labelling 1 mg of protein or 30 nmol of amino groups.
规格
Environmentally Preferable | ||
主机 |