Amersham™ CyDye™ Value Packs (Bis-Reactive NHS Esters)
供应商: Cytiva
PA15500 PA15502 PA15504 PA13000 PA13002 PA13004 PA15000 PA15002 PA15004 PA17000 PA17002 PA17004
GEHEPA15500EA 0 CNY
GEHEPA15500 GEHEPA15502 GEHEPA15504 GEHEPA13000 GEHEPA13002 GEHEPA13004 GEHEPA15000 GEHEPA15002 GEHEPA15004 GEHEPA17000 GEHEPA17002 GEHEPA17004
Amersham™ CyDye™ Value Packs (Bis-Reactive NHS Esters)
蛋白连接和标记系统
Amersham™ CyDye™ fluors combine exceptional photostability with bright and intense signals, resulting in high sensitivity and a highly reproducible performance at a biologically suitable pH range of 3 to 10.
  • Wide selection of CyDye fluors for protein labeling for use in ELISA and immunoprecipitation available
  • Labeling strategy based on protein
  • Medium to high-throughput fluorescence imaging
  • Ready-to-use labeled antibodies enable multiplexed assays
  • Suitable for general labeling of amine residues
  • Large range of bulk packs available for greatest economy
Fluors are versatile fluorophores designed for a broad range of applications. The use of multicolor CyDye fluors enables multiplexing without crosstalk and readily dissolve in aqueous buffers, which eliminates the need of organic solvents. The high purity and thorough QC testing of CyDye fluors generate high levels of chromophore and reactive dye content. Narrow excitation and emission bands result in discrete signals from each fluor, which, together with minimal cross-talk, contributes to high accuracy.
The packs contain fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups.
Delivery information: Example protocols included with each dye pack.
Packaging: Packaged in foil packs, each containing 5, 10 or 50 mg dried dye.
现在订购
规格
Environmentally Preferable
主机