Mini vertical electrophoresis system, miniVE

供应商: GE HEALTHCARE
80-6418-77
GEHE80-6418-77EA 0 CNY
GEHE80-6418-77
Mini vertical electrophoresis system, miniVE
电泳系统 垂直电泳系统
The simple, modular design of miniVE vertical electrophoresis system consolidates all the small parts for electrophoresis and blotting into one convenient unit. Uniform gel temperature is maintained with the tank-style lower buffer chamber. A full tank stabilises the temperature so that lanes are straight and bands are flat. The blot module will accept two gel/membrane stacks simultaneously. The lower buffer chamber is designed to hold two blot modules, allowing four gels to be blotted at once.

  • Perform 1-D and 2-D electrophoresis and electrotransfer in a single compact unit
  • Cast, run,
More Product Information
现在订购

规格

型号 miniVE
凝胶尺寸 宽度×高度 100×105 mm
Environmentally Preferable
推荐的运行电压 300 V
玻璃板尺寸(宽度×高度) 100×105 mm
垫片 1,0 mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR