Chromatography columns, pre-packed
供应商: GE HEALTHCARE
29321909 29315195 29321910
GEHE29321909EA 0 CNY
GEHE29321909 GEHE29315195 GEHE29321910
Chromatography columns, pre-packed
色谱柱
Prepacked validation columns are suitable for scale-down studies such as viral clearance.
  • High-quality packing
  • Excellent dimensions for scale-down studies
  • Prepacked with the most commonly used BioProcess resins
  • High consistency between columns in terms of efficiency and asymmetry
Results of Analysis certificate available for each individual column - includes column performance testing such as asymmetry and efficiency. Developed together with biopharmaceutical experts to ensure optimal function for scale-down studies such as viral clearance.
现在订购
规格
Environmentally Preferable
pH 范围 2 - 14
存储 2...8 °C, 20% ethanol