Chemiluminiscent imager, ImageQuant™ LAS 500
供应商: GE HEALTHCARE
29-0050-63
GEHE29-0050-63EA 0 CNY
GEHE29-0050-63
Chemiluminiscent imager, ImageQuant™ LAS 500
成像系统
ImageQuant™ LAS 500 is a cooled CCD imager for imaging of chemiluminescent Western blots, fluorescent protein and DNA gel stains, and white light imaging of colorimetric stains and markers.
  • Sensitivity: detect picogram levels of sample with chemiluminiscence
  • Automatic overlay function: simultaneously image chemiluminescent samples and coloured molecular weight markers
  • Ready-to-operate in less than 5 min with the rapid cooling camera
  • Compact integrated design works independently of a dedicated PC with flexible image storage
  • Flexible storage of image on a
More Product Information