Columns, PD MiniTrap™ G-10
供应商: GE HEALTHCARE
28-9180-10
GEHE28-9180-10EA 0 CNY
GEHE28-9180-10
Columns, PD MiniTrap™ G-10
色谱柱
PD MiniTrap G-10 columns are prepacked, single-use gravity columns for buffer exchange and cleanup of biological samples, for example peptides, small proteins, and oligosaccharides.
  • Easy cleanup of peptides, small proteins, or carbohydrates larger than Mr 700 before downstream analysis such as liquid chromatography, electrophoresis, and MS
  • Efficient removal of contaminants such as salts, dyes, and radioactive labels
  • High desalting capacity
  • Preparation of samples in the volume range of 100 to 300 μl
PD MiniTrap G-10 gravity columns give fast and simpleMore Product Information