Film, Amersham Hyperfilm™ MP
供应商: GE HEALTHCARE
28-9068-50 28-9068-46 28-9068-45 28-9068-44 28-9068-43 28-9068-42
GEHE28-9068-50EA 0 CNY
GEHE28-9068-50 GEHE28-9068-46 GEHE28-9068-45 GEHE28-9068-44 GEHE28-9068-43 GEHE28-9068-42
Film, Amersham Hyperfilm™ MP
放射自显影胶片 薄膜
Amersham Hyperfilm™ MP is a multipurpose film constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer. The anti-scratch coating reduces the risk of visible artefacts (e.g., plastic wrap marks) and the clear colourless base results in easy reading of images, without compromising on sensitivity.
  • High sensitivity for low concentration targets
  • Intended for radiometric or chemiluminescent applications
  • Clear background for excellent contrast and improved band visibility
  • Publication quality images
More Product Information