Ab buffer kit
供应商: Cytiva
28-9030-59
GEHE28-9030-59EA 0 CNY
GEHE28-9030-59
Ab buffer kit
缓冲(液) Antibody Purification Buffers
Ab buffer kit contains prepared buffer concentrates for binding, washing, and elution of IgG according to recommended protocols.
The kit also includes neutralising buffer. Ab buffer kit eliminates time consuming buffer preparation, which allows fast, reproducible, and convenient purification work.
Delivery information: Includes: 1×50 ml binding buffer (10X stock solution), 1×15 ml elution buffer (10X stock solution), and 1×25 ml neutralising buffer.