Anti-IgG Goat Polyclonal Antibody (Cy3®)
供应商: GE HEALTHCARE
28-9011-06
GEHE28-9011-06EA 0 CNY
GEHE28-9011-06
Anti-IgG Goat Polyclonal Antibody (Cy3®)
抗体
ECL Plex Cy-conjugated Antibodies are part of an optimized system for quantitative analysis using fluorescent western blotting.
Optimized system for quantitative analysis using fluorescent Western blotting. ECL Plex Western Blotting System is optimized for single protein detection as well as multiplex protein detection using ECL Plex CyDye-Conjugated Secondary Antibodies and Hybond-LFP (low-fluorescent PVDF membrane) or Hybond ECL (nitrocellulose membrane). The optimized antibodies show high sensitivity, a very good dynamic range and low or no cross-reactivity. CyDye technology isMore Product Information