Affinity chromatography media, MabSelect™ PrismA
供应商: Cytiva
17-5498-02
GEHE17-5498-02EA 0 CNY
GEHE17-5498-02
Affinity chromatography media, MabSelect™ PrismA
蛋白纯化系统 Protein Purification Resins
MabSelect™ PrismA has been improved with an optimised high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing future demands in mAb processing to be met.
  • Enhanced dynamic binding capacity allows high mass throughput of processed mAb per resin volume unit
  • Excellent alkaline stability enables efficient cleaning and sanitisation using 0,5 to 1,0 M NaOH for improved process economy, bioburden, control and robustness
  • Covered by a comprehensive security of supply program, including dual sources of the agarose base matrix and protein A ligand
MabSelect™ PrismA builds on the proven track record of MabSelect™ and MabSelect™ SuRe™ resins in commercial mAb production.
现在订购
规格
配体 Alkaline-stabilised protein A-derived (E.coli)
Environmentally Preferable
配体偶联法 Epoxy
基质 Rigid, highly cross-linked agarose
pH 范围 3 – 12
结合量 ~ 40 mg polyclonal IgG/ml resin, 2 min residence time
~ 80 mg polyclonal IgG/ml resin, 6 min residence time
存储温度 2 - 8 °C