Hydrophobic interaction chromatography columns, SOURCE™ 15PHE PE
供应商: GE HEALTHCARE
17-5186-01
GEHE17-5186-01EA 0 CNY
GEHE17-5186-01
Hydrophobic interaction chromatography columns, SOURCE™ 15PHE PE
色谱柱
SOURCE 15PHE 4.6/100 PE is a Tricorn high performance column pre-packed with SOURCE 15PHE.
  • High resolution separations at high loads and high flow rates
  • Wide range of selectivities
  • Reproducible quality
  • Excellent scalability
  • High chemical and physical stability
  • Low backpressure
SOURCE 15PHE 4.6/100 PE high capacity makes it an excellent choice for preparative hydrophobic interaction purifications at laboratory scale. In addition, its longer bed height provides increased resolution. The column is also useful for optimization studies during scale-up.