Percoll™
供应商: GE HEALTHCARE
17-0891-01 17-0891-02 17-0891-09
GEHE17-0891-01EA 0 CNY
GEHE17-0891-01 GEHE17-0891-02 GEHE17-0891-09
Percoll™
细胞分离介质
Percoll® is commonly used for separating cells, organelles, viruses, and other subcellular particles.
  • For Research Use Only
  • Commonly used in basic research applications
  • Used in balanced salt solutions
  • Colorless to slightly yellow
A silica-based density gradient media for use in separating cells, organelles, viruses, and other subcellular particles. Percoll® comprises silica particles coated with polyvinylpyrrolidone (PVP) and is commonly used in basic research applications. Also used in balanced salt solutions, physiological saline or 0,25 M sucrose,More Product Information