Sepharose™ CL-6B Gel Filtration Base Matrix
供应商: Cytiva
17-0160-01
GEHE17-0160-01EA 0 CNY
GEHE17-0160-01
Sepharose™ CL-6B Gel Filtration Base Matrix
蛋白纯化系统 Protein Purification Resins
Sepharose™ CL-6B is a cross-linked agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix.
  • 6% cross-linked agarose gel filtration media
  • Proven base matrix for coupling affinity ligands
  • Higher degree of crosslinking improve matrix stability
The matrix is not pre-activated and the user performs all steps in coupling.
现在订购
规格
说明 Sepharose™ CL-6B
Environmentally Preferable
基质 Cross-linked agarose, 6%, spherical
流速 <30 cm/h
分数化范围 1 × 104 - 4 × 106 globular proteins
pH 范围 3 - 13
存储温度 4 - 30 °C