Restriction enzymes, HindIII, FastDigest™, Fermentas

供应商: Thermo Fisher Scientific

FD0505
FERMFD0505EA 0 CNY
FERMFD0505
Restriction enzymes, HindIII, FastDigest™, Fermentas
核酸试剂 Restriction Enzymes

Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.


  • 100% activity of all FastDigest™ enzymes in one universal reaction buffer
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications


Designed to complete digestion in 5 to 15 minutes.

现在订购

规格

Environmentally Preferable
Application
Enzyme name HindIII
Source
Overhang type 5' overhang
Compatible buffer FastDigest Green Buffer or FastDigest Buffer (colorless)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR