Bottles, narrow neck, round, Boston type
供应商: EP SCIENTIFIC
S349-0125 149-0500 149-1000 249-0500 249-1000
EPSCS349-0125EA 0 CNY
EPSCS349-0125 EPSC149-0500 EPSC149-1000 EPSC249-0500 EPSC249-1000
Bottles, narrow neck, round, Boston type
玻璃瓶
Soda lime glass, Class III, amber
Ideal for solvent, collecting water samples for a variety of organic and inorganic target analytes. Amber coloured for light-sensitive materials. These bottles are available as certified, processed and unprocessed with the following features.
Certified bottles: It is designed to meet or exceed EPA Performance based specifications for semi-volatile organics, pesticides, PCBs and metals analyses. Each bottle is individually barcoded with lot number and unique container number for traceability and certificate of analysis (CofA) is included in the case.
现在订购
规格
Environmentally Preferable Low Manufacturing Impact
环境认证 NAR Certificate of Retirement