Rotors for Centrifuge 5425/5425 R
供应商: EPPENDORF
5495505.008 5495510.001
EPPE5495505.008EA 0 CNY
EPPE5495505.008 EPPE5495510.001
Rotors for Centrifuge 5425/5425 R
离心机 转子
Eppendorf rotors for centrifuge 5425/5425 R, precision meets versatility. Optimise your centrifuge with a range of capacities and speeds. Explore accessory options such as adaptors and buckets for tailored sample processing, ensuring optimal results in molecular biology and beyond.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customise your centrifuge setup
  • Aerosol-tight and PTFE coated options available
More Product Information