Eppendorf Xplorer® Electronic Multi-Channel Pipettes
供应商: EPPENDORF
4861000.120 4861000.163 4861000.171
EPPE4861000.120EA 0 CNY
EPPE4861000.120 EPPE4861000.163 EPPE4861000.171
Eppendorf Xplorer® Electronic Multi-Channel Pipettes
移液管和移液器 移液器
Eppendorf Xplorer® multi-channel pipettes with dispensing mode provide a cutting-edge solution for high-throughput liquid handling. Offering the highest levels of simplicity, precision, and reproducibility, these pipettes will help you pipette with more speed, comfort, and ergonomics.
  • Electronic single, 8 and 12 channel pipettes designed to aspirate and dispense liquid volumes from 0,5 μl to 10 ml
  • Intuitive operating concept and ergonomic design
  • Multifunctional rocker and ergonomic display angle
  • Selection dial and innovative ejector
  • Convenient handle with
More Product Information