Eppendorf twin.tec® PCR Plates 96-well, 250 µl
供应商: EPPENDORF
0030128.575 0030128.605 0030128.869 0030129.326 0030129.334 0030133.366 0030133.374 0030133.390 0030133.404 0030129.610
EPPE0030128.575EA 0 CNY
EPPE0030128.575 EPPE0030128.605 EPPE0030128.869 EPPE0030129.326 EPPE0030129.334 EPPE0030133.366 EPPE0030133.374 EPPE0030133.390 EPPE0030133.404 EPPE0030129.610
Eppendorf twin.tec® PCR Plates 96-well, 250 µl
微孔板 PCR板
Eppendorf's twin.tec® 96-well, 250 µl PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid and torque-resistant polycarbonate
More Product Information