Eppendorf twin.tec® PCR Plates 384-Well, 45 µl
供应商: EPPENDORF
0030128.508 0030128.532 0030128.931 0030128.966 0030129.342 0030129.350 0030129.628
EPPE0030128.508EA 0 CNY
EPPE0030128.508 EPPE0030128.532 EPPE0030128.931 EPPE0030128.966 EPPE0030129.342 EPPE0030129.350 EPPE0030129.628
Eppendorf twin.tec® PCR Plates 384-Well, 45 µl
微孔板 PCR板
Eppendorf's twin.tec® 384-well, 45 µl PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimising the risk of cross-contamination and contributing to consistently reliable results.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid and torque-resistant polycarbonate
More Product Information