epT.I.P.S.® 384 Reloads - Micropipette Tips
供应商: EPPENDORF
0030076.001 0030076.010 0030076.044 0030076.052
EPPE0030076.001EA 0 CNY
EPPE0030076.001 EPPE0030076.010 EPPE0030076.044 EPPE0030076.052
epT.I.P.S.® 384 Reloads - Micropipette Tips
吸头
Eppendorf epT.I.P.S.® 384 Reloads provide high-precision micropipette tips in 'Eppendorf Quality' and 'PCR clean' grades, tailored for 384-well plate applications, ensuring accuracy, reduced waste, and compatibility in high-throughput labs. The epT.I.P.S.® 384 pipette tips build a system together with the Research® plus and Xplorer® plus 16- and 24-channel pipettes, as well as 20 and 100 µl Move It® adjustable tip spacing pipettes for the utmost safety and reliability.
  • Designed for a cone distance of 4,5 mm
  • Innovative SOFTattach technology allows for
More Product Information