Anti-PTPRC Mouse Monoclonal Antibody (Alexa Fluor® 532) [clone: HI30]
供应商: EBIOSCIENCE
58-0459-42
EBIO58-0459-42EA 0 CNY
EBIO58-0459-42
Anti-PTPRC Mouse Monoclonal Antibody (Alexa Fluor® 532) [clone: HI30]
抗体
Description: The HI30 monoclonal antibody reacts with all isoforms of human CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding circulating erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes. Applications Reported: This HI30 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HI30 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells.More Product Information