Anti-IFNgamma Rat Antibody (eFluor® 450) [clone: XMG1.2]

供应商: EBIOSCIENCE
48-7311-82
EBIO48-7311-82EA 0 CNY
EBIO48-7311-82
Anti-IFNgamma Rat Antibody (eFluor® 450) [clone: XMG1.2]
抗体
Type:
Antigen: IFNgamma
Clonality:
Clone: XMG1.2
Conjugation: eFluor® 450
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR