Gas detection tube for hydrogen sulphide
供应商: Dräger
CH29101 8101831 8101211
DRAECH29101EA 0 CNY
DRAECH29101 DRAE8101831 DRAE8101211
Gas detection tube for hydrogen sulphide
充气管
To measure the 'spot concentrations' of hydrogen sulphide for use with an Accuro® pump. Uses include determination and measurement of concentration peaks, measurement of personal exposure in the inhalation area, detection of leaks in, for example, pipelines or gas installations, and air analysis in sewers, shafts and confined spaces.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20 °C and 1013 hPa.
Operating conditions:
0,2/a: 10 to 30 °C ; 3 to 15 mg/l H₂O absolute humidity
0,2/b: 15 to 40 °C ; Up to 2More Product Information