Microplate stands
供应商: DIVERSIFIED BIOTECH
WPST-1000
DIVEWPST-1000EA 0 CNY
DIVEWPST-1000
Microplate stands
Well Plate Stands and Well Orienters
Convenient holder for microplates to comfortably view samples and pipette without strain. Available tilted at a 30° angle or as an adjustable version allowing the flexibility to adjust the angle of the microplate (45° max. tilt).