Identification cards, multi-coloured, 96-well and 384-well
供应商: DIVERSIFIED BIOTECH
WELL-96H WELL-96V
DIVEWELL-96HEA 0 CNY
DIVEWELL-96H DIVEWELL-96V
Identification cards, multi-coloured, 96-well and 384-well
Well Plate Stands and Well Orienters
Visual aid for pipetting and identification of samples in microplates. Fits most standard 96-well and 384-well plates.
  • Bright colours make it easy to distinguish between columns and rows
Can be used with the Pipette Guide for convenient viewing.