Silicone rubber stoppers, conical
供应商: DEUTSCH NEUMANN
1010503 1010505 1010508 1010510 1010512 1010514 1010517 1010518 1010521 1010523 1010526 1010529 1010531 1010536 1010541 1010547 1010550 1010556 1010560 1010564 1010571 1010579 1010587 1010594 1011510 1011512 1011514 1011517 1011518 1011521 1011523 1011526 1011529 1011531 1012514 1012517 1012529 1012526 1012518 1012521 1012523 1012531
DENE1010503EA 0 CNY
DENE1010503 DENE1010505 DENE1010508 DENE1010510 DENE1010512 DENE1010514 DENE1010517 DENE1010518 DENE1010521 DENE1010523 DENE1010526 DENE1010529 DENE1010531 DENE1010536 DENE1010541 DENE1010547 DENE1010550 DENE1010556 DENE1010560 DENE1010564 DENE1010571 DENE1010579 DENE1010587 DENE1010594 DENE1011510 DENE1011512 DENE1011514 DENE1011517 DENE1011518 DENE1011521 DENE1011523 DENE1011526 DENE1011529 DENE1011531 DENE1012514 DENE1012517 DENE1012529 DENE1012526 DENE1012518 DENE1012521 DENE1012523 DENE1012531
Silicone rubber stoppers, conical
塞子 硅胶瓶塞
These stoppers are biologically inert and transparent.
  • Resistant to temperatures from −60 to +220 °C and for short periods up to +250 °C
Certifications: DIN 12 871