Disposable particle filtering respirators, FFP1/FFP2/FFP3, 4000 series
供应商: HONEYWELL SAFETY PRODUCTS
1005605 1005608 1005611 1005614 1005630
DALL1005605EA 0 CNY
DALL1005605 DALL1005608 DALL1005611 DALL1005614 DALL1005630
Disposable particle filtering respirators, FFP1/FFP2/FFP3, 4000 series
呼吸器 一次性防护口罩 Particulate Respirators
Individually wrapped fold-flat respirators with the exclusive features of the Willson 5000 series: ultra-soft seal, new exhalation valve, latex, PVC, silicone-free materials.
  • Five models in a fold-flat style presented in hygienic individual sachets printed with user instructions
  • Can be conveniently stored in a pocket
  • Fold and hold mechanism prevents contaminants from entering the masks when not in use
  • Chin piece specifically designed to allow users normal movement during speech
EN149:2001
Certifications: EN 149:2001