Disposable particle filtering respirators, FFP1/FFP2/FFP3, 5000 series
供应商: HONEYWELL SAFETY PRODUCTS
1005580 1005113 1005582 1005586-V2 1005099 1005602-V2 1005588 1005593 1005591 1005595 1005598 1005600
DALL1005580EA 0 CNY
DALL1005580 DALL1005113 DALL1005582 DALL1005586-V2 DALL1005099 DALL1005602-V2 DALL1005588 DALL1005593 DALL1005591 DALL1005595 DALL1005598 DALL1005600
Disposable particle filtering respirators, FFP1/FFP2/FFP3, 5000 series
呼吸器 一次性防护口罩 Particulate Respirators
  • Ultra-soft seal providing a soft textile sensation on the skin, absorbing perspiration, ensuring a good seal; hypoallergenic qualities
  • 'Butterfly' exhalation valve offering maximum frontal protection on the valve membrane while also providing minimal exhalation resistance
  • Latex-free, PVC-free and silicone-free materials
  • Dolomite-tested: Test demonstrating the respirator's resistance to clogging over time; ensures better breathing comfort and less worker fatigue
Colour codes: FFP1D - yellow, FFP2D - green, FFP3D - orange, FFP1D-OV - brown for organicMore Product Information