Anti-Rb Rabbit Monoclonal Antibody [clone: D20]

供应商: CELL SIGNALING TECHNOLOGIES
9313S
CSTA9313SEA 0 CNY
CSTA9313S
Anti-Rb Rabbit Monoclonal Antibody [clone: D20]
抗体
Type: Primary
Antigen: Rb
Clonality: Monoclonal
Clone: D20
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Monkey
现在订购

规格

抗原 Rb
Environmentally Preferable
抗体类型 Primary
克隆形成能力 Monoclonal
克隆 D20
反应性 Monkey,Human,Mouse
主机 Rabbit
同位型
蛋白质印迹 Yes
免疫沉淀反应法 Yes
ChIP Yes
顺序 CENILKRVKDVGHIFKEKFANAVGQG
存储温度 –20 °C
装运温度 Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR