Anti-Lysine ubiquitinate K48 linked (ubiquitin) Rabbit Polyclonal Antibody

供应商: CELL SIGNALING TECHNOLOGIES
4289S
CSTA4289SEA 0 CNY
CSTA4289S
Anti-Lysine ubiquitinate K48 linked (ubiquitin) Rabbit Polyclonal Antibody
抗体
Type: Primary Antibodies
Antigen: Lysine ubiquitinate K48 linked (ubiquitin)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Hoat: Rabbit
Isotype:
Reactivity: All
现在订购

规格

抗原 Lysine ubiquitinate K48 linked (ubiquitin)
Environmentally Preferable
抗体类型 Primary
克隆形成能力 Polyclonal
反应性 Multiple
主机 Rabbit
同位型
蛋白质印迹 Yes
顺序 CXLXFXGK(GGRLR)QLEXX
存储温度 −20 °C
装运温度 Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR