Anti-Met Mouse Monoclonal Antibody [clone: L41G3]

供应商: CELL SIGNALING TECHNOLOGIES
3148S
CSTA3148SEA 0 CNY
CSTA3148S
Anti-Met Mouse Monoclonal Antibody [clone: L41G3]
抗体
Type: Primary
Antigen: Met
Clonality: Monoclonal
Clone: L41G3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
现在订购

规格

抗原 Met
Environmentally Preferable
抗体类型 Primary
克隆形成能力 Monoclonal
克隆 L41G3
反应性 Monkey,Human
主机 Mouse
同位型 IgG1
蛋白质印迹 Yes
免疫沉淀反应法 Yes
顺序 VAPYPSLLSSEDNADDEVDTRPASFWETSC
存储温度 −20 °C
装运温度 Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR