Anti-p53 Rabbit Monoclonal Antibody [clone: 7F5]

供应商: CELL SIGNALING TECHNOLOGIES
2527S
CSTA2527SEA 0 CNY
CSTA2527S
Anti-p53 Rabbit Monoclonal Antibody [clone: 7F5]
抗体
Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: 7F5
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Monkey
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR