Overhead stirrer shafts
供应商: COWIE
005.440.2 005.450.2 005.455.2 005.465.2 005.240.2 005.250.2 005.255.2 005.265.2 005.340.2 005.350.2 005.355.2 005.365.2 005.355.1.2 005.365.1.2 005.0265.12.2 005.0465.12.2 005.255.10.2 005.430.8.2 005.440.8.2 005.450.8.2 005.455.10.2 005.0465.10.2 005.0475.10.2 005.0475.12.2 005.04100.12.2 005.330.8.2 005.340.8.2 005.350.8.2 005.355.10.2 005.0365.10.2 005.0375.10.2 005.0365.12.2 005.0375.12.2 005.03100.12.2 005.230.8.2 005.240.8.2 005.250.8.2 005.0265.10.2 005.0275.10.2 005.0275.12.2 005.02100.12.2 005.0475.16.2 005.0465.2 005.04100.16.2 005.0375.16.2 005.0375.1.2 005.0365.1.2 005.03100.16.2 005.0275.2 005.230.2 005.02100.16.2 005.430.2 005.0275.16.2 005.0475.2 005.330.2 005.0265.2
COWI005.440.2EA 0 CNY
COWI005.440.2 COWI005.450.2 COWI005.455.2 COWI005.465.2 COWI005.240.2 COWI005.250.2 COWI005.255.2 COWI005.265.2 COWI005.340.2 COWI005.350.2 COWI005.355.2 COWI005.365.2 COWI005.355.1.2 COWI005.365.1.2 COWI005.0265.12.2 COWI005.0465.12.2 COWI005.255.10.2 COWI005.430.8.2 COWI005.440.8.2 COWI005.450.8.2 COWI005.455.10.2 COWI005.0465.10.2 COWI005.0475.10.2 COWI005.0475.12.2 COWI005.04100.12.2 COWI005.330.8.2 COWI005.340.8.2 COWI005.350.8.2 COWI005.355.10.2 COWI005.0365.10.2 COWI005.0375.10.2 COWI005.0365.12.2 COWI005.0375.12.2 COWI005.03100.12.2 COWI005.230.8.2 COWI005.240.8.2 COWI005.250.8.2 COWI005.0265.10.2 COWI005.0275.10.2 COWI005.0275.12.2 COWI005.02100.12.2 COWI005.0475.16.2 COWI005.0465.2 COWI005.04100.16.2 COWI005.0375.16.2 COWI005.0375.1.2 COWI005.0365.1.2 COWI005.03100.16.2 COWI005.0275.2 COWI005.230.2 COWI005.02100.16.2 COWI005.430.2 COWI005.0275.16.2 COWI005.0475.2 COWI005.330.2 COWI005.0265.2
Overhead stirrer shafts
搅拌器 Stirring Tools
PTFE, with encapsulated steel reinforced drive shaft. Chemically inert. The smooth surface reduces contamination and makes cleaning easier.
  • Non scratch, prevents agitation vessels from being damaged
  • Temperature resistant from −200 to +280 °C
Caution: EX denotes exposed end, these stirrers have a stainless steel shaft core with an exposed end for rigid clamping.