Corning® Disposable Phenolic Threaded Screw Cap with Rubber Liners, Corning
供应商: CORNING SCIENCE PRODUCTS
99999-15
CORN99999-15EA 0 CNY
CORN99999-15
Corning® Disposable Phenolic Threaded Screw Cap with Rubber Liners, Corning
外壳 检测/样品管盖
These disposable Corning® caps are made from phenolic resin and are autoclavable at 121°C
  • Available in different sizes
They are provided with a white rubber liner.
现在订购
规格
Environmentally Preferable Low Manufacturing Impact
环境认证 I-RECs, ISO 14001 - Monterrey