External thread cryogenic vials
供应商: CORNING SCIENCE PRODUCTS
430658 430659 430661 430662 430663
CORN430658EA 0 CNY
CORN430658 CORN430659 CORN430661 CORN430662 CORN430663
External thread cryogenic vials
小瓶
These vials are manufactured from polypropylene to withstand temperatures down to –196 °C.
  • Black graduations with large marking spot
  • Vials have a silicone washer for a secure seal
  • 2 ml self-standing vials are available in five cap colors.
  • Vials may be color coded with inserts
  • Free foam rack with each case
  • RNase-/DNase-free
  • Sterile
  • Nonpyrogenic
Self-standing vials have a special base design allowing them to be locked into cryogenic rack and tray (Cat. No. 431131) for single-handed manipulation.
Caution: Do not use cryogenic vials for storage in theMore Product Information