Corning® Treated Culture Dishes
供应商: CORNING SCIENCE PRODUCTS
430165 430166 430167 430196 430293 430599 431110 3261 3262 3295
CORN430165EA 0 CNY
CORN430165 CORN430166 CORN430167 CORN430196 CORN430293 CORN430599 CORN431110 CORN3261 CORN3262 CORN3295
Corning® Treated Culture Dishes
培养皿 组织培养皿
Optically clear PS. Available with a choice of treated surface. Corning® CellBIND® surface increases surface wetability for more even and consistent cell attachment. Ultra-Low Attachment dishes feature a covalently bound hydrogel layer that minimises cell attachment, protein absorption and cellular activation.
  • With stacking beads to aid handling
  • Supplied with vents to provide consistent gas exchange
  • Sterilised by gamma radiation
  • Certified non-pyrogenic
These disposable petri-style dishes are manufactured from optically clear virgin polystyrene. DishesMore Product Information