Clear inserts, polyester (PET) membrane, Transwell®
供应商: CORNING SCIENCE PRODUCTS
3450 3452 3460 3462 3470 3472 3464
CORN3450EA 0 CNY
CORN3450 CORN3452 CORN3460 CORN3462 CORN3470 CORN3472 CORN3464
Clear inserts, polyester (PET) membrane, Transwell®
微孔板 插入板和插入物
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
  • w Produce a cell culture environment that closely resembles an in vivo state
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 µm transparent membrane
  • Treated for optimal cell attachment
  • Excellent visibility under phase contrast microscopy
  • Supplied in multiple well plates
  • Sterile
Allow polarised cells to carryMore Product Information
现在订购
规格
Environmentally Preferable Low Manufacturing Impact
环境认证 RECs, ISO 14001 - Kennebunk