Cell culture chambers, Corning® CellSTACK®
供应商: CORNING SCIENCE PRODUCTS
3268 3269 3270 3271 3272 3319 3310 3320 3321 3330 3311 3312 3303 3313
CORN3268EA 0 CNY
CORN3268 CORN3269 CORN3270 CORN3271 CORN3272 CORN3319 CORN3310 CORN3320 CORN3321 CORN3330 CORN3311 CORN3312 CORN3303 CORN3313
Cell culture chambers, Corning® CellSTACK®
生物反应器
PS, sterile. The Corning® CellSTACK® culture chambers are available in five sizes and with three different surface treatments.
  • Two 26 mm diameter filling ports allow direct access to chamber bottom providing greater flexibility for sterile filling and emptying by pouring, pipetting or via tubing in a fully closed system
  • Standard 33 mm threaded caps have 0,2 μm pore non-wettable membranes sealed directly to the caps to allow gas exchange while minimising the risk of contamination
  • Optional 33 mm threaded caps are available with integrally sealed USP Class VI
More Product Information