Culture system, Corning® E-Cube™
供应商: CORNING SCIENCE PRODUCTS
3286
CORN3286EA 0 CNY
CORN3286
Culture system, Corning® E-Cube™
生物反应器
The E-Cube™ System is a simple bioreactor with 8500 cm² cell growth area for growing anchorage dependent cells in only a 25,4×35,6 cm footprint. The E-Cube™ System provides a simple method to determine if cells will grow in the CellCube® Module prior to investing in the resources and funding that would be necessary for the larger, automated CellCube® System. Peristaltic pump, CO₂ incubator and medium are not provided.
Includes an oxygenator, medium reservoir, multiple access ports, and all required tubing and fittings.
Cells grow in Corning’'s parallel-plate CellCube® Module on theMore Product Information