Media/storage bottles, narrow mouth, round, with screw cap, PYREX®
供应商: CORNING SCIENCE PRODUCTS
1395-100 1395-25 1395-250 1395-50 1395-500 1395250HTC
CORN1395-100EA 0 CNY
CORN1395-100 CORN1395-25 CORN1395-250 CORN1395-50 CORN1395-500 CORN1395250HTC
Media/storage bottles, narrow mouth, round, with screw cap, PYREX®
玻璃瓶
Heavy duty bottles, made of PYREX® borosilicate glass, with orange PP linerless, one-piece autoclavable plug seal screw with a drip-free pouring ring.
  • Permanent white enamel graduations with marking spots
  • Glass bead indicates full capacity line
  • Very good temperature resistance, very good chemical resistance
Graduated, are used for storage as well as mixing and sampling.
Further colour-coded PP caps, PBT caps, ETFE pouring rings (for dry heat sterilisation (180 °C)), and vented membrane caps are available.
Caution: Do not tighten cap immediatelyMore Product Information