Cell culture vessels, Corning® HYPERflask™ M, CellBIND® treated
供应商: CORNING SCIENCE PRODUCTS
10024 10020 10030 10034
CORN10024EA 0 CNY
CORN10024 CORN10020 CORN10030 CORN10034
Cell culture vessels, Corning® HYPERflask™ M, CellBIND® treated
烧瓶 细胞培养瓶
Optically clear polystyrene flasks feature a straight neck, flat-top cap, with or without bar code for use manually or with automated systems. No refrigeration or special handling required.
These exceptionally high-yield, high-performance flasks utilise a multilayered gas permeable growing surface for efficient gas exchange. Unique design allows flasks to produce 10× higher yields than standard flasks, increasing productivity and capacity as well as reducing processing time and incubator space. Ten interconnected polystyrene surfaces provide 1720 cm² area for cell growth with an air gapMore Product Information